1,3-Dihydroxy-2-(hydroxymethyl)anthracene-9,10-dione 98%

Supplier: Ambeed
Danger

Synonyms: Lucidin

A602082-10MG A602082-1MG A602082-25MG A602082-50MG A602082-5MG
76805-972EA 112.46 USD
76805-972 76805-974 76805-976 76805-978 76805-980
1,3-Dihydroxy-2-(hydroxymethyl)anthracene-9,10-dione 98%
1,3-Dihydroxy-2-(hydroxymethyl)anthracene-9,10-dione

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₅
MW: 270.23 g/mol
MDL Number: MFCD01664482
CAS Number: 478-08-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR