H-Val-Trp-OH 99+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((S)-2-Amino-3-methylbutanamido)-3-(1H-indol-3-yl)propanoic acid

A599894-100MG A599894-1G A599894-250MG A599894-25G A599894-5G
76805-960EA 11.67 USD
76805-960 76805-962 76805-964 76805-966 76805-968
H-Val-Trp-OH 99+%
H-Val-Trp-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₁N₃O₃
MW: 303.36 g/mol
MDL Number: MFCD00037947
CAS Number: 24587-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR