6-Methoxyquinoline N-Oxide 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxyquinoline 1-oxide

A596345-10G A596345-1G A596345-250MG A596345-5G
76805-944EA 250.38 USD
76805-944 76805-946 76805-948 76805-950
6-Methoxyquinoline N-Oxide 97%
6-Methoxyquinoline N-Oxide

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00009771
CAS Number: 6563-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR