2-Bromo-1-(1-methyl-1H-imidazol-2-yl)ethanone 95%

Supplier: Ambeed
Danger

A595571-100MG A595571-250MG
76805-940EA 39.26 USD
76805-940 76805-942
2-Bromo-1-(1-methyl-1H-imidazol-2-yl)ethanone 95%
2-Bromo-1-(1-methyl-1H-imidazol-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.03 g/mol
MDL Number: MFCD11656633
CAS Number: 750556-81-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR