Ethyl 3-hydrazino-3-oxopropionate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-hydrazinyl-3-oxopropanoate

A594078-100G A594078-10G A594078-1G A594078-25G A594078-5G
76805-932EA 496.51 USD
76805-932 76805-934 76805-936 76805-938 77669-566
Ethyl 3-hydrazino-3-oxopropionate 96%
Ethyl 3-hydrazino-3-oxopropionate

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₃
MW: 146.145 g/mol
MDL Number: MFCD00134484
CAS Number: 30866-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR