Diethylene glycol dibenzoate 90%

Supplier: Ambeed
Warning

Synonyms: Oxybis(ethane-2,1-diyl) dibenzoate

A591033-1000G A591033-100G A591033-25G A591033-500G
76805-898EA 75.33 USD
76805-898 76805-900 76805-902 76805-904
Diethylene glycol dibenzoate 90%
Diethylene glycol dibenzoate

Precaución: for Research Use Only

Formula: C₁₈H₁₈O₅
MW: 314.34 g/mol
MDL Number: MFCD00020679
CAS Number: 120-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR