3-(Diethylcarbamoyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(N,N-Diethylaminocarbonyl)phenylboronic acid, 3-(Diethylcarbamoyl)benzeneboronic acid, (3-(Diethylcarbamoyl)phenyl)boronic acid

A590215-1G A590215-250MG A590215-5G
76805-892EA 44.56 USD
76805-892 76805-894 76805-896
3-(Diethylcarbamoyl)phenylboronic acid 97%
3-(Diethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD04115684
CAS Number: 237413-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR