Methyl (methylsulfinyl)methyl sulfide 98%

Supplier: Ambeed
Danger

Synonyms: FAMSO, Methyl((methylsulphinyl)methyl)sulphane, Methyl (Methylthio)methyl sulphoxide, Formaldehyde Dimethyl Dithioacetal S-Oxide

A589620-1G A589620-25G A589620-5G
76805-884EA 6 USD
76805-884 76805-886 76805-888
Methyl (methylsulfinyl)methyl sulfide 98%
Methyl (methylsulfinyl)methyl sulfide

Precaución: for Research Use Only

Formula: C₃H₈OS₂
MDL Number: MFCD00002091
CAS Number: 33577-16-1
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR