2-Aminobutanamide 97%

Supplier: Ambeed
Danger

Synonyms: H-Abu-NH₂, (S)-2-Aminobutanamide

A589141-1G A589141-25G A589141-5G
76805-878EA 23.34 USD
76805-878 76805-880 76805-882
2-Aminobutanamide 97%
2-Aminobutanamide

Precaución: for Research Use Only

Formula: C₄H₁₀N₂O
MW: 102.137 g/mol
MDL Number: MFCD00136552
CAS Number: 7324-11-0
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR