Ethyl 2-propylpentanoate 95%

Supplier: Ambeed
Warning

A587118-1G A587118-250MG
76805-852EA 56 USD
76805-852 76805-854
Ethyl 2-propylpentanoate 95%
Ethyl 2-propylpentanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MDL Number: MFCD00870991
CAS Number: 17022-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR