2-Methylpropanohydrazide 98%

Supplier: Ambeed
Danger

Synonyms: Isobutyrohydrazide, Isobutyric acid Hydrazide

A586441-1G A586441-5G
76805-848EA 11 USD
76805-848 76805-850
2-Methylpropanohydrazide 98%
2-Methylpropanohydrazide

Precaución: for Research Use Only

Formula: C₄H₁₀N₂O
MDL Number: MFCD00025122
CAS Number: 3619-17-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR