(S)-2-Aminosuccinic acid, sodium salt 98%

Supplier: Ambeed
Warning

A584251-1000G A584251-100G A584251-500G
76805-826EA 27.59 USD
76805-826 76805-828 76805-830
(S)-2-Aminosuccinic acid, sodium salt 98%
(S)-2-Aminosuccinic acid, sodium salt

Precaución: for Research Use Only

Formula: C₄H₇NNaO₄
MW: 156.09 g/mol
MDL Number: MFCD00037209
CAS Number: 3792-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR