1-(Cyclohexylmethyl)piperazine 95%

Supplier: Ambeed
Warning

A583162-100G A583162-250MG A583162-5G
76805-820EA 1294.3 USD
76805-820 76805-822 76805-824
1-(Cyclohexylmethyl)piperazine 95%
1-(Cyclohexylmethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂
MW: 182.311 g/mol
MDL Number: MFCD01075237
CAS Number: 57184-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR