N-Ethyl-N-((triethoxysilyl)methyl)ethanamine 95+%

Supplier: Ambeed
Danger

A583024-100G A583024-500G A583024-25G
76805-816EA 42.44 USD
76805-816 76805-818 77627-204
N-Ethyl-N-((triethoxysilyl)methyl)ethanamine 95+%
N-Ethyl-N-((triethoxysilyl)methyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₁H₂₇NO₃Si
MW: 249.42 g/mol
MDL Number: MFCD01728991
CAS Number: 15180-47-9
UN: 3267
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR