5-Chlorobenzo[b]thiophene-3-acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid

A582475-100MG A582475-250MG
76805-808EA 62.6 USD
76805-808 76805-810
5-Chlorobenzo[b]thiophene-3-acetic acid 98%
5-Chlorobenzo[b]thiophene-3-acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₇ClO₂S
MW: 226.68 g/mol
MDL Number: MFCD00052308
CAS Number: 17266-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR