[2-(4-Aminomethylphenoxy)ethyl]dimethylamine 97%

Supplier: Ambeed
Warning

A581845-100G A581845-1G A581845-25G A581845-5G A581845-10G
76805-800EA 454.99 USD
76805-800 76805-802 76805-804 76805-806 77669-528
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine 97%
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD01075231
CAS Number: 20059-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR