Ethyl benzimidate hydrochloride 97%

Supplier: Ambeed
Warning

A577921-10G A577921-25G A577921-5G A577921-100G
76805-778EA 32.89 USD
76805-778 76805-780 76805-782 77602-434
Ethyl benzimidate hydrochloride 97%
Ethyl benzimidate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MDL Number: MFCD00043246
CAS Number: 5333-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR