Ethyl-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-4-carboxylate

A577817-1G A577817-250MG A577817-5G
76805-774EA 71.09 USD
76805-774 76805-776 77669-520
Ethyl-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate 97%
Ethyl-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂S
MW: 226.3 g/mol
MDL Number: MFCD08689777
CAS Number: 76263-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR