Ethyl-p-tolylacetate 98%

Supplier: Ambeed
Warning

A576536-100G A576536-10G A576536-1G A576536-25G A576536-5G
76805-754EA 149 USD
76805-754 76805-756 76805-758 76805-760 76805-762
Ethyl-p-tolylacetate 98%
Ethyl-p-tolylacetate

Precaución: for Research Use Only

Formula: (C₆H₄(CH₃))CH₂COOCH₂CH₃
MDL Number: MFCD00009181
CAS Number: 14062-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR