Methotrexate hydrate 98%

Supplier: Ambeed
Danger

A575327-1G A575327-25G A575327-5G
76805-746EA 15 USD
76805-746 77602-416 77602-418
Methotrexate hydrate 98%
Methotrexate hydrate

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₈O₅·xH₂O
MDL Number: MFCD00150847
CAS Number: 133073-73-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR