Isopropyl palmitate 97%

Supplier: Ambeed
Warning

A573195-100G A573195-25G A573195-500G A573195-1000G
76805-730EA 10 USD
76805-730 76805-732 76805-734 77602-412
Isopropyl palmitate 97%
Isopropyl palmitate

Precaución: for Research Use Only

Formula: C₁₉H₃₈O₂
MDL Number: MFCD00008993
CAS Number: 142-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR