N-(4-Bromophenyl)maleimide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Bromophenyl)-1H-pyrrole-2,5-dione
A571621-1G
A571621-5G
76805-716EA
14
USD
76805-716
77880-230
N-(4-Bromophenyl)maleimide 98%
N-(4-Bromophenyl)maleimide
Precaución: for Research Use Only
Formula:
C₁₀H₆BrNO₂ MW: 252.07 g/mol |
MDL Number:
MFCD00030664 CAS Number: 13380-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...