Nicardipine hydrochloride 99%

Supplier: Ambeed
Danger

Synonyms: 3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, Nicardipine HCl

A570738-1G A570738-1MG A570738-250MG A570738-25G
76805-710EA 33 USD
76805-710 76805-712 76805-714 77669-508
Nicardipine hydrochloride 99%
Nicardipine hydrochloride

Precaución: for Research Use Only

Formula: C₂₆H₂₉N₃O₆
MW: 515.99 g/mol
MDL Number: MFCD00057327
CAS Number: 54527-84-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR