3-Ethyl-2,5-dimethylpyrazine 97%

Supplier: Ambeed
Warning

A569451-100MG A569451-50MG A569451-1G A569451-250MG
76805-700EA 122 USD
76805-700 77572-812 77880-226 77880-227
3-Ethyl-2,5-dimethylpyrazine 97%
3-Ethyl-2,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD00053098
CAS Number: 13360-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR