Ethyl-2,4-dichlorobenzoate 98+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,4-dichlorobenzoate

A568822-1G A568822-25G A568822-5G
76805-694EA 18.04 USD
76805-694 76805-696 76805-698
Ethyl-2,4-dichlorobenzoate 98+%
Ethyl-2,4-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
MDL Number: MFCD00277702
CAS Number: 56882-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR