Potassium carboxyformate 98%

Supplier: Ambeed
Warning

A568473-1000G A568473-100G A568473-500G
76805-690EA 21 USD
76805-690 76805-692 77880-225
Potassium carboxyformate 98%
Potassium carboxyformate

Precaución: for Research Use Only

Formula: C₂HKO₄
MDL Number: MFCD00036407
CAS Number: 127-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR