Ethyl linoleate 70%

Supplier: Ambeed
Warning

Synonyms: 9Z,12Z-Octadecadienoic acid ethyl ester, Linoleic acid ethyl ester, cis,cis-9,12-Octadecadienoic acid ethyl ester, (9Z,12Z)-Ethyl octadeca-9,12-dienoate

A567680-100G A567680-25G A567680-500G
76805-680EA 65.78 USD
76805-680 76805-682 76805-684
Ethyl linoleate 70%
Ethyl linoleate

Precaución: for Research Use Only

Formula: C₂₀H₃₆O₂
MW: 308.5 g/mol
MDL Number: MFCD00009535
CAS Number: 544-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR