Imidazo[1,2-a]pyridin-6-ylmethanol 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Hydroxymethyl)imidazo[1,2-a]pyridine

A565843-1G A565843-250MG
76805-670EA 78.51 USD
76805-670 76805-672
Imidazo[1,2-a]pyridin-6-ylmethanol 97%
Imidazo[1,2-a]pyridin-6-ylmethanol

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.1600 g/mol
MDL Number: MFCD07368521
CAS Number: 132213-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR