2-Hydroxyterephthalic acid dimethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 2-hydroxyterephthalate

A562172-1G A562172-250MG A562172-25G A562172-5G
76805-626EA 44.56 USD
76805-626 76805-628 76805-630 76805-632
2-Hydroxyterephthalic acid dimethyl ester 95%
2-Hydroxyterephthalic acid dimethyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
MDL Number: MFCD08445666
CAS Number: 6342-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR