Hexyl benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Benzoic acid hexyl ester

A561237-1000G A561237-100G A561237-25G A561237-500G
76805-618EA 95.49 USD
76805-618 76805-620 76805-622 76805-624
Hexyl benzoate 98%
Hexyl benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
MDL Number: MFCD00051714
CAS Number: 6789-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR