Ethyl-4-fluorobenzoylformate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-fluorophenyl)-2-oxoacetate

A560433-25G A560433-5G A560433-10G
76805-616EA 172 USD
76805-616 77669-484 77880-204
Ethyl-4-fluorobenzoylformate 98%
Ethyl 4-fluorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
MDL Number: MFCD01319617
CAS Number: 1813-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR