Diethyl (2-bromoethyl)phosphonate 98%

Supplier: Ambeed
Warning

A560002-100G A560002-1G A560002-5G
76805-608EA 134 USD
76805-608 76805-610 77572-770
Diethyl (2-bromoethyl)phosphonate 98%
Diethyl (2-bromoethyl)phosphonate

Precaución: for Research Use Only

Formula: C₆H₁₄BrO₃P
MDL Number: MFCD00000239
CAS Number: 5324-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR