Ethyl-O-tolylacetate 98%

Supplier: Ambeed
Warning

A559083-1G A559083-25G A559083-5G
76805-592EA 11 USD
76805-592 76805-594 76805-596
Ethyl-O-tolylacetate 98%
Ethyl-O-tolylacetate

Precaución: for Research Use Only

Formula: (C₆H₄CH₃)CH₂COOCH₂CH₃
MDL Number: MFCD00009179
CAS Number: 40291-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR