Iron(II) phthalocyanine 98%

Supplier: Ambeed
Danger

Synonyms: Phthalocyanine Iron(II), Iron phthalocyanine

A559064-5G A559064-25G
76805-590EA 6.37 USD
76805-590 77602-378
Iron(II) phthalocyanine 98%
Iron(II) phthalocyanine

Precaución: for Research Use Only

Formula: C₃₂H₁₆FeN₈
MW: 568.38 g/mol
MDL Number: MFCD00015953
CAS Number: 132-16-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR