2,4,5-Triphenylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: Lophine, 2,4,5-Triphenyl-1H-imidazole

A554550-100G A554550-10G A554550-1G A554550-5G
76805-530EA 195.21 USD
76805-530 76805-532 76805-534 76805-536
2,4,5-Triphenylimidazole 98%
2,4,5-Triphenylimidazole

Precaución: for Research Use Only

Formula: C₂₁H₁₆N₂
MW: 296.37 g/mol
MDL Number: MFCD00005187
CAS Number: 484-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR