Ethyl-6-fluoro-4-hydroxyquinoline-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate
A554005-1G
A554005-250MG
A554005-25G
A554005-5G
76805-522EA
24.41
USD
76805-522
76805-524
76805-526
76805-528
Ethyl-6-fluoro-4-hydroxyquinoline-3-carboxylate 98%
Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₀FNO₃ MW: 235.22 g/mol |
MDL Number:
MFCD00173345 CAS Number: 318-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...