SIA (N-Succinimidyl iodoacetate) 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 2-iodoacetate, N-Succinimidyl iodoacetate, Iodoacetic acid N-succinimidyl Ester, N-(Iodoacetoxy)succinimide, SIA

A553805-1MG A553805-1G A553805-250MG A553805-100MG A553805-5G
76805-520EA 5 USD
76805-520 77602-346 77602-348 77669-446 77669-448
SIA (N-Succinimidyl iodoacetate) 95%
SIA (N-Succinimidyl iodoacetate)

Precaución: for Research Use Only

Formula: C₆H₆INO₄
MDL Number: MFCD00058451
CAS Number: 39028-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR