Ethyl-4-(4-chlorophenyl)-2,4-dioxobutanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-(4-chlorophenyl)-2,4-dioxobutanoate

A552387-100MG A552387-1G A552387-250MG A552387-5G
76805-506EA 29.71 USD
76805-506 76805-508 76805-510 76805-512
Ethyl-4-(4-chlorophenyl)-2,4-dioxobutanoate 98%
Ethyl-4-(4-chlorophenyl)-2,4-dioxobutanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClO₄
MW: 254.67 g/mol
MDL Number: MFCD01935972
CAS Number: 5814-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR