Glycerine 99.5%

Supplier: Ambeed
Warning

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

A551901-1000G A551901-100G A551901-25G A551901-500G
76805-498EA 15 USD
76805-498 76805-500 76805-502 76805-504
Glycerine 99.5%
Glycerine

Precaución: for Research Use Only

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Flash Pt: 177
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR