2,8-Dimethylquinolin-4(1H)-one 98%

Supplier: Ambeed
Danger

A551281-1G A551281-25G A551281-5G
76805-486EA 10 USD
76805-486 76805-488 76805-490
2,8-Dimethylquinolin-4(1H)-one 98%
2,8-Dimethylquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MDL Number: MFCD00100264
CAS Number: 15644-80-1
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR