4-(Pyrimidin-5-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

A551068-1G A551068-250MG A551068-5G
76805-480EA 39 USD
76805-480 76805-482 76805-484
4-(Pyrimidin-5-yl)benzaldehyde 98%
4-(Pyrimidin-5-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O
MDL Number: MFCD02684108
CAS Number: 198084-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR