Vinyldiphenylphosphine 98% stabilized

Supplier: Ambeed
Warning

Synonyms: Diphenyl(vinyl)phosphine

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A549243-1G A549243-25G A549243-5G
76805-462EA 32.89 USD
76805-462 76805-464 76805-466
Vinyldiphenylphosphine 98% stabilized
Vinyldiphenylphosphine

Precaución: for Research Use Only

Formula: C₁₄H₁₃P
MW: 212.2268 g/mol
MDL Number: MFCD00008611
CAS Number: 2155-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR