Fmoc-D-Tle-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-tBu-D-Gly-OH

A547654-1G A547654-25G A547654-5G A547654-100G
76805-426EA 66 USD
76805-426 76805-428 76805-430 77880-183
Fmoc-D-Tle-OH 98%
Fmoc-D-Tle-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
MDL Number: MFCD00065650
CAS Number: 198543-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR