DIPSO 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
A547600-500G
A547600-25G
A547600-100G
A547600-10G
A547600-5G
76805-424EA
203.7
USD
76805-424
77602-312
77627-128
77627-130
77627-132
DIPSO 95%
DIPSO
Precaución: for Research Use Only
Formula:
C₇H₁₇NO₆S MW: 261.29 g/mol |
MDL Number:
MFCD00038353 CAS Number: 68399-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...