Diphemanil Methylsulfate 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Diphenylmethylene)-1,1-dimethylpiperidin-1-ium methyl sulphate, 4-(Diphenylmethylene)-1,1-dimethylpiperidin-1-ium Methyl sulphate, Diphenmanil methylsulphate

A547556-10MG A547556-1MG A547556-50MG
76805-418EA 14.86 USD
76805-418 76805-420 76805-422
Diphemanil Methylsulfate 98%
Diphemanil Methylsulfate

Precaución: for Research Use Only

Formula: C₂₁H₂₇NO₄OS
MW: 405.51 g/mol
MDL Number: MFCD00865157
CAS Number: 62-97-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR