Methyl-4-chlorocinnamate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(4-chlorophenyl)acrylate, 4-Chlorocinnamic Acid Methyl Ester

A544663-25G A544663-5G
76805-402EA 37.14 USD
76805-402 76805-404
Methyl-4-chlorocinnamate 98%
Methyl-4-chlorocinnamate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₂
MW: 196.63 g/mol
MDL Number: MFCD00075790
CAS Number: 7560-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR