Droperidol 98%

Supplier: Ambeed
Warning

Synonyms: 1-[1-[3-(p-Fluorobenzoyl)propyl]-1,2,3,6-tetrahydro-4-pyridyl]-2-benzimidazolinone, 1-(1-(4-(4-Fluorophenyl)-4-oxobutyl)-1,2,3,6-tetrahydropyridin-4-yl)-1H-benzo[d]imidazol-2(3H)-one

A541340-100MG A541340-1MG A541340-25MG A541340-1G A541340-250MG A541340-5G
76805-380EA 53.05 USD
76805-380 76805-382 76805-384 77627-108 77627-110 77627-112
Droperidol 98%
Droperidol

Precaución: for Research Use Only

Formula: C₂₂H₂₂FN₃O₂
MW: 379.43 g/mol
MDL Number: MFCD00083290
CAS Number: 548-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR