3,5-Dimethyl-4-propoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A539319-1G A539319-25G A539319-5G
76805-372EA 21.22 USD
76805-372 76805-374 76805-376
3,5-Dimethyl-4-propoxyphenylboronic acid 98%
3,5-Dimethyl-4-propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
MW: 208.0619 g/mol
MDL Number: MFCD05865178
CAS Number: 357611-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR