(±)-γ-Octalactone 98%

Supplier: Ambeed
Warning

Synonyms: (±)-Octan-4-olide, 5-Butyldihydrofuran-2(3H)-one

A538934-1000G A538934-100G A538934-25G A538934-500G A538934-5G
76805-360EA 143.23 USD
76805-360 76805-362 76805-364 76805-366 76805-368
(±)-γ-Octalactone 98%
(±)-γ-Octalactone

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
MDL Number: MFCD00005402
CAS Number: 104-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR