exo-2-Aminonorbornane 95%

Supplier: Ambeed
Danger

Synonyms: (1R,2R,4S)-Bicyclo[2.2.1]heptan-2-amine, (1R,2R,4S)-rel-Bicyclo[2.2.1]heptan-2-amine, (1alpha,4alpha)-Norbornane-2alpha-amine

A538396-250MG A538396-5G A538396-1G A538396-25G
76805-346EA 71.09 USD
76805-346 76805-348 77551-630 77669-386
exo-2-Aminonorbornane 95%
exo-2-Aminonorbornane

Precaución: for Research Use Only

Formula: C₇H₁₃N
MW: 111.19 g/mol
MDL Number: MFCD00078132
CAS Number: 7242-92-4
UN: 2733
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR